Learn More
Fisherbrand™ Shell Vial Kit, Clear Glass, PE Plug
Recommended for HPLC usage.
Brand:  Fisherbrand™ 11 14 0544
13.11 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 11555914
  • 15.90€ / Pack of 100
In Stock
Description
  • Shell vials and the appropriate plugs can also be supplied as a 2in1 KIT
  • Handy and inexpensive vial/ closure combination for uncritical analyses
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Shell Vial
Glass
11
Clear
2.3 mL
No
SDS
Documents Certificates
Special Offers
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    18
    Hours
    19
    Minutes
    25
    Seconds
    40
  • Multiple Manufacturers
    Explore our new promotion and get up to 70% off some of our most popular products.
    Promo Code: 24127
    Expires2025-05-31
    Days
    49
    Hours
    19
    Minutes
    25
    Seconds
    40
Product Content Correction
Product Title
Fisherbrand™ Shell Vial Kit, Clear Glass, PE Plug > flat bottom,2ml,31,5mm height