Learn More
Endecotts™ Sieve Calibration Ball
Sieve balls, used to improve the sieving of cohesive material
Brand: Endecotts™ ZSICSA-.125
This item is not returnable.
View return policy
Description
- Spheres are traceable to the United Kingdom National Physical Laboratory
- Calibrate sieves to an accuracy of 1μm
- Soda-lime glass microspheres pass over the entire surface of the sieve and through every aperture
- Standards include a calibration chart
Specifications
Microspheres | |
Soda-lime glass | |
Sieve | |
120 mesh |