Learn More
Endecotts™ Sieve Calibration Ball
Sieve balls, used to improve the sieving of cohesive material
241.00€ - 414.00€
Specifications
Product Type | Microspheres |
---|---|
Material | Soda-lime glass |
For Use With (Equipment) | Sieve |
Product Code | Brand | Particle Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Price | Quantity & Availability | |||||
11924315
|
Endecotts™
ZSICSA-1.40 |
14 mesh |
414.00€
Pack of 3 |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||||
11994295
|
Endecotts™
ZSICSA-.125 |
120 mesh |
241.00€
Pack of 5 |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||||
11934295
|
Endecotts™
ZSICSA-.038 |
400 mesh |
252.00€
Pack of 5 |
Estimated Shipment: 28-11-2024
Log in to see stock. |
|||||
Description
- Spheres are traceable to the United Kingdom National Physical Laboratory
- Calibrate sieves to an accuracy of 1μm
- Soda-lime glass microspheres pass over the entire surface of the sieve and through every aperture
- Standards include a calibration chart
Specifications
Microspheres | |
Soda-lime glass | |
Sieve |