Learn More
Greiner Bio-One Small Volume™ 384 Deep Well Polypropylene Microplates
Ensures accurate pipetting, reduces dead volume. Greiner Bio-One Small Volume™ 384 Deep Well Polypropylene Microplates are specifically designed to accommodate heat-sealing systems.
Brand:  Greiner Bio-One 784209
 View more versions of this product
Product Code. 11993276
  • 1035.00€ / Pack of 100
Estimated Shipment: 25-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Non-pyrogenic
  • Conical well design with flat bottom
  • Ensures accurate pipetting
  • Reduces dead volume of well
  • Square well geometry, compatible with CapMat plate sealers
  • Accommodates heat-sealing systems
  • Service Plan
    No
    Services
    No
    TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
Black
384
1 to 90 μL
16 x 24
Polypropylene
Non-sterile
Deep well