Learn More
Greiner Bio-One Small Volume™ 384 Deep Well Polypropylene Microplates
Ensures accurate pipetting, reduces dead volume. Greiner Bio-One Small Volume™ 384 Deep Well Polypropylene Microplates are specifically designed to accommodate heat-sealing systems.
1035.00€
Specifications
Format | Standard |
---|---|
Array | 16 x 24 |
Material | Polypropylene |
No. of Wells | 384 |
Well Design | Deep well |
Description
- Non-pyrogenic
- Conical well design with flat bottom
- Ensures accurate pipetting
- Reduces dead volume of well
- Square well geometry, compatible with CapMat plate sealers
- Accommodates heat-sealing systems
-
Service Plan
No
Warranty
No
Specifications
Standard | |
Polypropylene | |
Deep well |
16 x 24 | |
384 |