Learn More
Greiner Bio-One Small Volume™ 384 Deep Well Polypropylene Microplates
Ensures accurate pipetting, reduces dead volume. Greiner Bio-One Small Volume™ 384 Deep Well Polypropylene Microplates are specifically designed to accommodate heat-sealing systems.
1035.00€
Specifications
Format Standard
Array 16 x 24
Material Polypropylene
No. of Wells 384
Well Design Deep well
View More Specs
Products 1
Product Code Brand Sterility Color Price Quantity & Availability  
Product Code Brand Sterility Color Price Quantity & Availability  
11993276
View Documents Promotion Details
Greiner Bio-One
784209
Non-sterile Black
1035.00€
Pack of 100
Estimated Shipment: 25-11-2024
Description
  • Non-pyrogenic
  • Conical well design with flat bottom
  • Ensures accurate pipetting
  • Reduces dead volume of well
  • Square well geometry, compatible with CapMat plate sealers
  • Accommodates heat-sealing systems
  • Service Plan
    No
    Warranty
    No
Specifications
Specifications
Standard
Polypropylene
Deep well
16 x 24
384