Learn More
DWK Life Sciences Kimble Chase™ Soda Lime Glass Culture Test Tubes
Use Soda Glass Cell Culture Test Tubes to analyze cell culture material that are critical to diagnoses and experimentation.
Brand: DWK Life Sciences A407512000611
11.78 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Soda lime glass
- Round bottom
- X250
Specifications
Round-bottom | |
75 mm | |
250 Pack | |
12 mm |
Test Tube | |
Soda Lime Glass | |
Autoclavable |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%