Learn More
Cytiva Spin Adapters for Gravity Columns
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
15.00€ - 20.00€
Specifications
Description | Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns. |
---|---|
Material | Polypropylene |
Quantity | 10 Pack |
Type | Spin Column Adapter |
Product Code | Brand | Column Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Column Type | Price | Quantity & Availability | ||||||
11717829
|
Cytiva
28-9232-45 |
For PD-10 Desalting Column |
20.00€
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11707829
|
Cytiva
28-9232-44 |
For PD MidiTrap G-25 Column |
20.00€
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11797819
|
Cytiva
28-9232-43 |
For PD MiniTrap G-25 Column |
15.00€
Pack of 10 |
In Stock
Log in to see stock. |
||||||
Description
- Convenient adapters, enable spinning of all gravity columns.
Specifications
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns. | |
Polypropylene | |
10 Pack | |
Spin Column Adapter |
Product Title
Cytiva Spin Adapters for Gravity Columns