Learn More
OHAUS™ Splashing Shield
High Quality Accessories for Hot Plate Stirrers
66.00€ - 83.00€
Specifications
For Use With (Application) | For Guardian 5000 range |
---|---|
Quantity | 1 |
Type | Hot Plate Kit |
Material | Plastic |
Product Code | Brand | Model | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | ||||||
30182881
![]() |
OHAUS™
30910731 |
Splashing Shield for e-G52XX07C |
70.00€
1 unit |
Estimated Shipment: 20-06-2025
Log in to see stock. |
||||||
30182891
![]() |
OHAUS™
30910732 |
Splashing Shield for e-G52HS10C |
83.00€
1 unit |
Estimated Shipment: 20-06-2025
Log in to see stock. |
||||||
30182882
![]() |
OHAUS™
30910733 |
Splashing Shield for e-G52HSRDA |
66.00€
1 unit |
Estimated Shipment: 20-06-2025
Log in to see stock. |
||||||
Description
- Accessory for Guardian™ Hotplates and Stirrers
Specifications
For Guardian 5000 range | |
1 | |
Hot Plate Kit | |
Plastic |
Product Title
OHAUS™ Splashing Shield