Learn More
Brady™ StainerBondz™ Microscope Slide Matte Polyester Labels
Easily create chemical-resistant microscope slide labels with the StainerBondz™ polyester label material (B-481). Print almost anywhere with a Brady™ handheld label printer.
Marca: Brady™ 170884
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Available in presized shapes for commonly used slides and tissue cassettes in which superior chemical resistance is required
- Withstands the rigors of typical deparaffinization including ethanol and xylene, as well as hematoxylin and eosin slide staining processes
- Made with a permanent acrylic adhesive for tight bonding onto smooth surfaces in extreme conditions
Especificaciones
Autoclavable | |
M5-97-481 | |
None | |
Matte | |
Polyester | |
Thermal Transfer | |
Non-sterile | |
BMP51, M511 | |
-112°F to +266°F | |
0.9 x 0.9 in. | |
0.9 in. | |
0.5 in. |
RoHS Compliant to 2011/65/EU (Pass 2015/863) | |
Permanent Acrylic | |
Black on White | |
Die-cut | |
Top | |
Labels | |
Healthcare and Hospital Labeling, General Identification, Laboratory Labeling | |
280 Labels/Cartridge | |
0.0033 in. | |
0.9 in. | |
2.3 cm |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido