Learn More
Brady™ StainerBondz™ Microscope Slide Matte Polyester Labels
Easily create chemical-resistant microscope slide labels with the StainerBondz™ polyester label material (B-481). Print almost anywhere with a Brady™ handheld label printer.
54.91€ - 61.60€
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | RoHS Compliant to 2011/65/EU (Pass 2015/863) |
Adhesive Type | Permanent Acrylic |
Coating | None |
Color | Black on White |
Product Code | Brand | Dimensions (LxW) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (LxW) | Price | Quantity & Availability | |||||
17821789
|
Brady™
175660 |
0.25 x 1.05 in. |
61.60€
Pack of 360 |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
17871719
|
Brady™
170885 |
0.375 x 1 in. |
54.91€
Pack of 320 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
17811799
|
Brady™
170886 |
0.75 x 0.9 in. |
58.58€
Pack of 320 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
17851759
|
Brady™
170884 |
0.9 x 0.9 in. |
55.00€
Pack of 280 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
Description
- Available in presized shapes for commonly used slides and tissue cassettes in which superior chemical resistance is required
- Withstands the rigors of typical deparaffinization including ethanol and xylene, as well as hematoxylin and eosin slide staining processes
- Made with a permanent acrylic adhesive for tight bonding onto smooth surfaces in extreme conditions
Specifications
Autoclavable | |
Permanent Acrylic | |
Black on White | |
Die-cut | |
Top | |
Labels | |
Healthcare and Hospital Labeling, General Identification, Laboratory Labeling | |
-112°F to +266°F | |
0.5 in. |
RoHS Compliant to 2011/65/EU (Pass 2015/863) | |
None | |
Matte | |
Polyester | |
Thermal Transfer | |
Non-sterile | |
BMP51, M511 | |
0.0033 in. |