Learn More
Brady™ StainerBondz™ Polyester Labels for M611 and BMP61
Durable Polyester (B-490) markers are designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
Brand:  Brady™ 150880
 View more versions of this product
Product Code. 17100431
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Description
Description
  • Super chemical-resistant polyester (B-481) is designed for use in the slide staining process and other laboratory ID which requires superior chemical resistance.
  • Performs well in the commonly used H&E (hematoxilin and eosin) slide staining process.
Specifications
Specifications
Permanent Acrylic
Matte
Polyester
Inkjet
Non-sterile
-196°C
-320°F to +230°F
0.0032 in.
18.42 mm
41.28 mm
15.4 mm
9.53 mm
White
Roll
Single-side Printable
Labels
BMP™61 Label Printer; BradyPrinter M611 Mobile Label Printer
250 Labels/Roll
-196°C to +110°C
0.081 mm
1.625 in.
0.6 in.
41.28 mm
110 g