Learn More
Sampling Systems™ Stainless Steel Bucket
Sampling Systems™ Stainless Steel Buckets are made from a heavy gauge 316L Stainless Steel for durability.
Brand:  Sampling Systems™ A586-15
Additional Details : Weight : 20.16000kg
 View more versions of this product
Product Code. 15829701
  • 267.00€ / Each
Estimated Shipment: 12-12-2024
Description
  • Made from 316L Stainless Steel
  • Crevice free interior
  • Swing handle
  • Flat base