Learn More
Sampling Systems™ Stainless Steel Bucket
Sampling Systems™ Stainless Steel Buckets are made from a heavy gauge 316L Stainless Steel for durability.
226.00€ - 267.00€
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15809701
|
Sampling Systems™
A586-10 |
Capacity: 10L |
226.00€
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||||
15819701
|
Sampling Systems™
A586-12 |
Capacity: 12L |
244.00€
Each |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
15829701
|
Sampling Systems™
A586-15 |
Capacity: 15L |
267.00€
Each |
Estimated Shipment: 17-12-2024
Log in to see stock. |
|||||
Description
- Made from 316L Stainless Steel
- Crevice free interior
- Swing handle
- Flat base