Learn More
Linker Industrietechnik™ Stainless Steel Test Sieves
Constructed with a soft solder seam, and a sieve of stainless steel cloth.
Brand:  Linker Industrietechnik™ 552.025.222-051
 View more versions of this product
Product Code. 11749259
  • 108.00€ / Each
Only null left
Estimated Shipment: 08-08-2025
Description
  • Stainless steel ring construction
  • Stainless steel sieve cloth/skin
  • Soft solder seam
  • Heat resistant
  • Available in a variety of sieve mesh sizes
Specifications
Specifications
Heat resistant up to 170°C
2.1 EN 10 204, ISO 3310-1
50 mm
800 μm
800 μm
Internal: 25 mm
Stainless Steel
Stainless Steel
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Linker Industrietechnik™ Stainless Steel Test Sieves > Pore Size: 800 μm