Learn More
Linker Industrietechnik™ Stainless Steel Test Sieves
Constructed with a soft solder seam, and a sieve of stainless steel cloth.
107.00€ - 123.00€
Specifications
Description | Heat resistant up to 170°C |
---|---|
Certifications/Compliance | 2.1 EN 10 204, ISO 3310-1 |
Height (Metric) | Internal: 25 mm |
Frame Material | Stainless Steel |
Mesh Material | Stainless Steel |
Product Code | Brand | Pore Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | ||||||
11774369
|
Linker Industrietechnik™
550.025.222-022 |
63 μm |
112.00€
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
||||||
11799249
|
Linker Industrietechnik™
552.025.222-035 |
200 μm |
108.00€
Each |
Estimated Shipment: 02-05-2025
Log in to see stock. |
||||||
12644556
|
Linker Industrietechnik™
550.025.222032 |
150 μm |
123.00€
Each |
Estimated Shipment: 18-06-2025
Log in to see stock. |
||||||
11719259
|
Linker Industrietechnik™
552.025.222-043 |
400 μm |
119.00€
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
||||||
11819110
|
Linker Industrietechnik™
550.025.222038 |
250 μm |
119.00€
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
11729259
|
Linker Industrietechnik™
552.025.222-046 |
500 μm |
107.00€
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
11749259
|
Linker Industrietechnik™
552.025.222-051 |
800 μm |
108.00€
Each |
Estimated Shipment: 18-06-2025
Log in to see stock. |
||||||
11769259
|
Linker Industrietechnik™
552.025.222-054 |
1 mm |
108.00€
Each |
Estimated Shipment: 18-06-2025
Log in to see stock. |
||||||
Description
- Stainless steel ring construction
- Stainless steel sieve cloth/skin
- Soft solder seam
- Heat resistant
- Available in a variety of sieve mesh sizes
Specifications
Heat resistant up to 170°C | |
Internal: 25 mm | |
Stainless Steel |
2.1 EN 10 204, ISO 3310-1 | |
Stainless Steel |
Product Title
Linker Industrietechnik™ Stainless Steel Test Sieves