Learn More
Cole-Parmer™ Stainless Steel Tweezers
Recommended for micro electronics and precision assembly
Brand: Cole-Parmer™ 07287-08
Description
Recommended for micro electronics and precision assembly
- Hand finished points for accuracy and excellent tip hardness
- Anti-glare finish reduces eye strain
- Anti-magnetic and acid resistant
Specifications
Angled | |
4.25 in. | |
10.8 cm | |
Tweezer | |
Straight |
Each | |
Stainless Steel | |
Tweezers | |
Ea. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Cole-Parmer™ Stainless Steel Tweezers > Length: 107.9mm, Angled