Learn More
Cole-Parmer™ Stainless Steel Tweezers
Recommended for micro electronics and precision assembly
Brand: Cole-Parmer™ 07287-06
13.94 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Recommended for micro electronics and precision assembly
- Hand finished points for accuracy and excellent tip hardness
- Anti-glare finish reduces eye strain
- Anti-magnetic and acid resistant
Specifications
Ultra Fine Sharp | |
4.25 in. | |
10.8 cm | |
Tweezer | |
Straight |
Each | |
Stainless Steel | |
Tweezers | |
Ea. |