Learn More
Koettermann™ Standard Cabinet for Acids and Bases, PP, 520x600x1920mm
Koettermann™ Standard Cabinet for Acids and Bases provides suitable storage to accommodate a variety of laboratory needs, but is especially suited for storing acids and bases.
4235.00€ - 5495.00€
Specifications
Dimensions (D x W x H) | 520 x 600 x 1920 mm |
---|---|
Product Type | Acids and Bases Cabinet |
For Use With (Application) | Acids and Bases |
No. of Doors | 2 |
Product Code | Brand | Door Style | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Door Style | Includes | Price | Quantity & Availability | ||||
14835361
|
Koettermann™
377-00003 |
Left-hinged | 4 Drawers, Fan |
5495.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Currently Unavailable: We have no information when this product will be available.
View alternative products
|
|||||||||
14825361
|
Koettermann™
377-00001 |
Left-hinged | 4 Drawers, No Fan |
4235.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Currently Unavailable: We have no information when this product will be available.
View alternative products
|
|||||||||
15518545
|
Koettermann™
377-00002 |
Right-hinged | 4 Drawers, No Fan |
4235.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Currently Unavailable: We have no information when this product will be available.
View alternative products
|
Description
- Tall, Acid/Base Cabinet
- Two doors
- Four drawers
Specifications
520 x 600 x 1920 mm | |
Acids and Bases Cabinet | |
Acids and Bases | |
2 |
Product Title
Koettermann™ Standard Cabinet for Acids and Bases, PP, 520x600x1920mm