Learn More
Carl Zeiss™ Stemi 305 EDU Microscope Set
Compact Greenough stereo microscope with 5:1 zoom for biological education, labs and production environments.
Brand: Carl Zeiss™ 435063-9010-100
Description
- This compact Greenough stereo microscope allows you to observe your samples as they are: three-dimensional and crisp in contrast and no preparation required.
- The Stemi 305 is an easy-to-use microscope, requires minimal setup, everything is integrated including long-living LED illumination for both reflected and transmitted light.
- Spot Illuminator K LED - Single spot for brightfield and darkfield epi-illumination, Thread M24x0.5 for polarizer, Height adjustable, inclinable, zoomable.
Specifications
50/60 Hz | |
Microscope Set | |
Choice of interchangeable optics, stand, illuminator, eyepiece reticles, gliding, ball/socket and rotating stages, ergo hand rest for stand K LAB, software and cameras | |
100/240 V | |
8X to 40X Basic System, 4X to 200X with Interchangeable Lenses |
Stemi 305 | |
LED | |
400 Lp/mm to 1.25 ÎĽm | |
Greenough System | |
Alignment tasks, hobby and private applications, mechanical engineering, wood and paper industry, aerospace industry |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
CE_15332098.pdf
File size: 323.5 kB
Product Content Correction
Product Title
Carl Zeiss™ Stemi 305 EDU Microscope Set >