Learn More
STEMCELL Technologies STEMtaq™ Hot Start DNA Polymerase Master Mix Kit
Hot Start DNA polymerase master mix and nuclease-free water for PCR reactions
Brand:  STEMCELL Technologies 79013
Encompass_Preferred
Product Code. 17139901
  • POA / 1 set
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
  • STEMtaq™ Hot Start DNA Polymerase Master Mix Kit, which includes Hot Start master mix and nuclease-free water, reliably and consistently amplifies a wide range of PCR templates in a convenient master mix format that allows for room temperature setup
  • The enzyme is bound to a proprietary antibody that blocks activity until the initial denaturation step (94 - 95°C for 2 minutes) of the PCR re-activates the enzyme
  • Hot Start PCR can in some cases eliminate or minimize primer-dimers and secondary products for some targets, and potentially improve yield
  • STEMtaq™ Hot Start DNA Polymerase Master Mix is a 2X concentrated, ready-to-use solution containing the Hot Start version of the STEMtaq™ DNA polymerase, dNTPs, MgCl2, and reaction buffer
  • STEMtaq™ Hot Start DNA polymerase has 5' to 3' exonuclease activity and produces PCR fragments with a 3' A overhang
Specifications
Specifications
100 Reactions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.