Learn More
BD Filcon™ Syringe-type, Sterile, 70 μm
Filcon™, Syringe-type, are disposable filters for separating cells and nuclei by suction or compression.
Brand: BD 340605
444.48 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Porous membrane - enables large quantities of cell suspensions to be filtered
- Microporous PE funnel - designed to pass the biological material while trapping waste particles
- Syringe-type for separating by suction or compression
- Sterile
Specifications
70 μm | |
Sterile | |
Sterile |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Product Title
BD Filcon™ Syringe-type, Sterile, 70 μm > Filcon™ Syringe-type; Sterile: Yes; 70 μm