missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Sterility Bottles
Glass sterility bottles available with a variety of media
67.90€ - 215.00€
Specifications
Certifications/Compliance | USP |
---|---|
Form | Liquid |
Manufacturing Origin | United States |
Packaging Type | Bottle |
Product Code | Brand | Quantity | Color | Media Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Color | Media Type | Price | Quantity & Availability | |||
11713573
|
BD
290662 |
300 mL | Clear | Fluid D |
215.00€
3 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11703573
|
BD
290652 |
300 mL | Light Yellow | Fluid A |
200.00€
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11713593
|
BD
299107 |
100 mL | Beige | Trypticase Soy Broth |
77.50€
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11723573
|
BD
290831 |
100 mL | Clear | Fluid D |
67.90€
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Transparent labels for easy viewing and identification of medium
- Labels are laminated acetate to resist alcohol or chlorine treatment for cleanroom use
- Prepared bottles for sterility testing and microbial limits
Wide-Mouth Bottles
- Openings are 48mm in diameter to accommodate bulky samples
Order Info
Supplied in packs of 10 bottles
Compliance
Conform to harmonized USP requirements.
Specifications
USP | |
Liquid | |
United States | |
Bottle |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Sterility Bottles