Learn More
Buerkle™ SteriPlast™ Micro Spatulas
BIOPHARMAGREENER_CHOICE
Use to sample, transfer or process small amounts of chemicals, powders, granulates, pastes, creams or liquids. Buerkle™ SterilPlast™ Micro Spatulas facilitates precise work with sturdy centre handle.
Brand:  Buerkle™ 5378-1032
Product Code. 15624114
  • 72.45€ / Pack of 100
In Stock
to see stock.
Edge
Description
  • Polystyrene
  • Production, assembly and packaging according to clean room class 7
  • Complies with EU food requirements and FDA regulations
  • Packaged individually and sterilized by gamma rays
Specifications
Specifications
Polystyrene
1 mL, 0.25 mL
Sterile
EU food regulations
Videos
  • Buerkle LaboPlast Micro Spatulas
    Buerkle LaboPlast Micro Spatulas
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Buerkle LaboPlast Micro Spatulas
    SDS
    Documents Certificates
    Certificates
    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
    5 results found
    Lot Number Certificate Type Date Catalog Number
    Lot Number244609 Certificate TypeCertificate of Compliance Date10/01/2025 Catalog Number5378-1032
    Lot Number243291 Certificate TypeCertificate of Compliance Date20/09/2024 Catalog Number5378-1032
    Lot Number242195 Certificate TypeCertificate of Compliance Date26/07/2024 Catalog Number5378-1032
    Lot Number240615 Certificate TypeCertificate of Compliance Date13/03/2024 Catalog Number5378-1032
    Lot Number233074 Certificate TypeCertificate of Compliance Date25/07/2023 Catalog Number5378-1032
    Product Title
    Buerkle™ SteriPlast™ Micro Spatulas > 1 mL/0.25 mL