Learn More
Heidolph Instruments Stirring Bar Sets
Stirring bars available in oval and cylindrical shape sets
Brand: Heidolph Instruments 509-53000-00
Description
Oval Stir Bars:
- Shape is similar to that of the flask and ensures complete mixing
- Equipped with an oval magnetic core, more force is available than with a cylindrical magnetic core
- For applications in:
- Round bottom flasks e.g. for reaction mixtures
Cylindrical Stir Bars:
- Smooth operating conditions at optimum contact area
- Long lifetime
- For applications in:
- Beakers
- Crystallizing dishes
- Heating bathes
- Other flat bottom vessels
Specifications
Autoclavable | |
15 x 6 mm | |
PTFE | |
15 mm | |
3/Pk. |
White | |
10 mL Flasks | |
6 mm | |
PTFE | |
Oval |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Heidolph Instruments Stirring Bar Sets > Size: 15 x 6 mm; Oval