missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ STX7 Recombinant Protein

Brand:  Abnova™ H00008417-P01.25ug

 View more versions of this product

Product Code. 16192432

  • 508.00€ / 25µg
Estimated Shipment: 03-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH11975
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
54.45
8
Glutathione Sepharose 4 Fast Flow
25 μg
MSYTPGVGGDPAQLAQRISSNIQKITQCSVEIQRTLNQLGTPQDSPELRQQLQQKQQYTNQLAKETDKYIKEFGSLPTTPSEQRQRKIQKDRLVAEFTTSLTNFQKVQRQAAEREKEFVARVRASSRVSGSFPEDSSKERNLVSWESQTQPQVQVQDEEITEDDLRLIHERESSIRQLEADIMDINEIFKDLGMMIHEQGDVIDSIEANVENAEVHVQQANQQLSRAADYQRKSRKTLCIIILILVIGVAIISLIIWGLNH
STX7
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
8417
STX7 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
STX7
Wheat Germ (in vitro)
GST
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers