missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TBX3 Recombinant Protein Antigen
Recombinant Protein
Brand: Novus Biologicals™ NBP3-21394PEP
This item is not returnable.
View return policy
Description

Specifications
Antibody Competition | |
6926 | |
100 μL | |
bladder cancer related protein XHL, T-box 3, T-box protein 3, T-box transcription factor TBX3, TBX3-ISO, ulnar mammary syndrome, UMS, XHL | |
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP3-21394. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL | |
>80% by SDS-PAGE and Coomassie blue staining |
PBS and 1 M Urea, pH 7.4. | |
MolecularWeight-theroretical: 28 kDa | |
Store at -20°C. Avoid freeze/thaw cycles. | |
TBX3 | |
Angiogenesis, Apoptosis, Cardiovascular Biology, Cell Biology | |
TBX3 |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ TBX3 Recombinant Protein Antigen > 100 μg
Spot an opportunity for improvement?Share a Content Correction