missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGD4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
383.00€ - 611.00€
Specifications
Antigen | TIGD4 |
---|---|
Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18392332
|
Bio-Techne
NBP3-17318-25UL |
25 μg |
383.00€
25µL |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
18355866
|
Bio-Techne
NBP3-17318-100UL |
100 μg |
611.00€
100µL |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
Description
TIGD4 Polyclonal antibody specifically detects TIGD4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
TIGD4 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
MGC43837, tIgGer transposable element derived 4, tIgGer transposable element-derived protein 4 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: MTSDVFEQWMRKLDEEFQAQQRRVVIFVESFPAHPEVKNLKSIELAFFPSCLSSKCIAMKQGVIKSLKIKYRHCLIKKFLSSVEGSKEF | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Purified | |
RUO | |
PBS, pH 7.2, 40% glycerol | |
201798 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title