Learn More
TOE1 Mouse, anti-Human, Unconjugated, Clone: 2C10H7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67829-1-IG-20UL
This item is not returnable.
View return policy
Description
The TOE1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human TOE1. This antibody recognizes human antigen. The TOE1 antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
TOE1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
2C10H7 | |
Unconjugated | |
20 μL | |
114034 | |