Learn More
Aalborg™ Tripod Base For Tripod Tube Frames
Aalborg™ Tripod Base For Tripod Tube Frames supports single and multi tripod tube frames to add stability.
Brand:  Aalborg™ TP2
 View more versions of this product
Product Code. 11738378
  • 83.00€ / Each
Estimated Shipment: 17-04-2025
Description
  • Accessory for P, S, and T style meter
  • For use with single or multi tube frame
  • Accommodates 65mm and 150mm flowmeter
Specifications
Specifications
Tripod 2 Multi Tube Frames
Tripod Base
Documents
One moment while we fetch your results.
Product Title
Aalborg™ Tripod Base For Tripod Tube Frames > For Use With: 65 and 150mm flowmeter