Learn More
Aalborg™ Tripod Base For Tripod Tube Frames
Aalborg™ Tripod Base For Tripod Tube Frames supports single and multi tripod tube frames to add stability.
83.00€ - 97.00€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11738378
|
Aalborg™
TP2 |
Tripod 2 Multi Tube Frames |
83.00€
Each |
Estimated Shipment: 17-03-2025
Log in to see stock. |
|||||
11758378
|
Aalborg™
TP1 |
Tripod Single Tube Frame |
83.00€
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||||
11767819
|
Aalborg™
TP5 |
Tripod 5 Multi Tube Frames |
97.00€
Each |
Estimated Shipment: 20-03-2025
Log in to see stock. |
|||||
11748378
|
Aalborg™
TP3 |
Tripod 3 Multi Tube Frames |
83.00€
Each |
Estimated Shipment: 20-03-2025
Log in to see stock. |
|||||
11787639
|
Aalborg™
TPG |
Tripod 2 Multi Tube Gas Frames |
83.00€
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||||
Description
- Accessory for P, S, and T style meter
- For use with single or multi tube frame
- Accommodates 65mm and 150mm flowmeter