Learn More
Aalborg™ Tripod Base For Tripod Tube Frames
Aalborg™ Tripod Base For Tripod Tube Frames supports single and multi tripod tube frames to add stability.
Brand: Aalborg™ TP1
This item is not returnable.
View return policy
Description
- Accessory for P, S, and T style meter
- For use with single or multi tube frame
- Accommodates 65mm and 150mm flowmeter
Specifications
Tripod Single Tube Frame | |
Tripod Base |
Product Content Correction
Product Title
Aalborg™ Tripod Base For Tripod Tube Frames > For Use With: 65 and 150mm flowmeter