Learn More
Aalborg™ Tripod Base For Tripod Tube Frames
Aalborg™ Tripod Base For Tripod Tube Frames supports single and multi tripod tube frames to add stability.
Brand:  Aalborg™ TP5
 View more versions of this product
Product Code. 11767819
  • 97.00€ / Each
Estimated Shipment: 24-04-2025
Description
  • Accessory for P, S, and T style meter
  • For use with single or multi tube frame
  • Accommodates 65mm and 150mm flowmeter
Specifications
Specifications
Tripod 5 Multi Tube Frames
Tripod Stand
Documents
One moment while we fetch your results.
Product Title
Aalborg™ Tripod Base For Tripod Tube Frames > For Use With: For 65 and 150mm flowmeter