Learn More
RETSCH Tungsten Carbide Grinding Jars
Use these grinding jars for dry, wet and cryogenic grinding of small samples. Retsch™ Tungsten Carbide Grinding Jars feature a self-centering mechanism and a screw-top design for leak-proof grinding.
Brand:  RETSCH 01.462.0217
 View more versions of this product
Product Code. 10541892
  • 2860.00€ / Each
Estimated Shipment: 19-02-2025
Description
  • Screw-top Tungsten grinding jars
  • Available in a variety of sizes
  • For use with Retsch mills
Specifications
Specifications
Grinding Jar
25 mL
For MM 301 Mixer mill
Tungsten Carbide