missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH Tungsten Carbide Grinding Jars
Use these grinding jars for dry, wet and cryogenic grinding of small samples. Retsch™ Tungsten Carbide Grinding Jars feature a self-centering mechanism and a screw-top design for leak-proof grinding.
934.00€ - 8275.00€
Product Code | Brand | Capacity (Metric) | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | For Use With (Equipment) | Price | Quantity & Availability | ||||
10541892
|
RETSCH
01.462.0217 |
25 mL | For MM 301 Mixer mill |
2860.00€
Each |
Estimated Shipment: 19-02-2025
Log in to see stock. |
||||
10212772
|
RETSCH
01.462.0235 |
10 mL | For MM 301 Mixer mill |
2630.00€
Each |
Estimated Shipment: 19-02-2025
Log in to see stock. |
||||
10394571
|
RETSCH
01.462.0009 |
10 mL | For MM 200 Mixer Mill | N/A | |||||
15310974
|
RETSCH
01.462.0317 |
50 mL | For EMAX |
8275.00€
Each |
Estimated Shipment: 19-02-2025
Log in to see stock. |
||||
10534041
|
RETSCH
01.462.0115 |
5 mL | For MM 200 Mixer Mill | N/A | |||||
Description
- Screw-top Tungsten grinding jars
- Available in a variety of sizes
- For use with Retsch mills
Specifications
Grinding Jar | |
Tungsten Carbide |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title