Learn More
Lenz Laborglasintrumente Vacuum Stopcocks, 1-Way, Parallel
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 02942910
 View more versions of this product
Product Code. 10570122
  • 153.00€ / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12545 and in addition to DIN standard
  • With parallel side arms and hollow glass plug
  • Material: borosilicate glass 3.3
  • Leakage rate acc. to DIN 12540-1
Specifications
Specifications
10 mm
Borosilicate Glass (3.3)