missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ Varian Split / Splitless Liner with Single Taper
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan Scientific and Medical™ 09203825
This item is not returnable.
View return policy
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Varian Bruker instruments
Specifications
25/Pk. | |
GC Inlet Liner, Split/Splitless Liner with Single Taper | |
5 mm |
54 mm | |
3.4 mm | |
Varian/Bruker GCs |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction