Learn More
Water, HPLC for Gradient Analysis, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber Glass Bottle
Plastic Coated Glass Bottle
Specifications
Specifications
Packaging | Amber Glass Bottle |
Quantity | 1 L |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2528146 | Certificate of Analysis | 24/02/2025 | W/0106/10RSS,W/0106/15X,W/0106/17X,W/0106/PB17,W/0106/17,W/0106/15,W/0106/17A,W/0106/25RSS |
2527436 | Certificate of Analysis | 17/02/2025 | W/0106/10RSS,W/0106/15X,W/0106/17X,W/0106/PB17,W/0106/17,W/0106/15,W/0106/17A,W/0106/25RSS |
2526366 | Certificate of Analysis | 11/02/2025 | W/0106/10RSS,W/0106/15X,W/0106/17X,W/0106/PB17,W/0106/17,W/0106/15,W/0106/17A,W/0106/25RSS |
2525710 | Certificate of Analysis | 06/02/2025 | W/0106/10RSS,W/0106/15X,W/0106/17X,W/0106/PB17,W/0106/17,W/0106/15,W/0106/17A,W/0106/25RSS |
Safety and Handling
Product Identifier
- Water
Special Offers
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Days
33Hours
13Minutes
18Seconds
21 -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
33Hours
13Minutes
18Seconds
21 -
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
3Hours
13Minutes
18Seconds
21
Product Content Correction
Product Title
Water, HPLC for Gradient Analysis, Fisher Chemical™ >