Learn More
YSI™ 605790 Pro Plus Quatro Cable
Measure conductivity/temperature, dissolved oxygen and any two ISEs among pH, ORP, ammonium, nitrate or chloride
Brand:  YSI™ 6057901
 View more versions of this product
Product Code. 11928089
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
  • A true field cable with MS (military spec) connectorsR
  • ugged, weighted probe guard
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Military Spec, Waterproof, Bayonet Lock
0.046 m
39.3 in.
1.85 in.
4-ports and 5560 temperature/conductivity sensor
1 m