Learn More
YSI™ 605790 Pro Plus Quatro Cable
Measure conductivity/temperature, dissolved oxygen and any two ISEs among pH, ORP, ammonium, nitrate or chloride
2233.80€ - 2284.80€
Specifications
Connector Types Military Spec, Waterproof, Bayonet Lock
Diameter (English) 1.85 in.
Diameter (Metric) 0.046 m
Includes 4-ports and 5560 temperature/conductivity sensor
View More Specs
Products 2
Product Code Brand Length (English) Length (Metric) Price Quantity & Availability  
Product Code Brand Length (English) Length (Metric) Price Quantity & Availability  
11928089
View Documents Promotion Details
YSI™
6057901
39.3 in. 1 m This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
11938089
View Documents Promotion Details
YSI™
6057904
157.4 in. 4 m This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
Description
Description
  • A true field cable with MS (military spec) connectorsR
  • ugged, weighted probe guard
Specifications
Specifications
Military Spec, Waterproof, Bayonet Lock
1.85 in.
0.046 m
4-ports and 5560 temperature/conductivity sensor