Learn More
YSI™ 605790 Pro Plus Quatro Cable
Measure conductivity/temperature, dissolved oxygen and any two ISEs among pH, ORP, ammonium, nitrate or chloride
Brand: YSI™ 6057904
This item is not returnable.
View return policy
Description
- A true field cable with MS (military spec) connectorsR
- ugged, weighted probe guard
Specifications
Military Spec, Waterproof, Bayonet Lock | |
0.046 m | |
157.4 in. | |
ProPlus handheld instrument |
1.85 in. | |
4-ports and 5560 temperature/conductivity sensor | |
4 m |