missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAY 678
Potent and selective cell-permeable human neutrophil elastase (HNE) inhibitor
118.00€ - 455.00€
Specifications
Chemical Name or Material | 5-[(4R)-5-Acetyl-1,2,3,4-tetrahydro-6-methyl-2-oxo-1-[3-(trifluoromethyl)phenyl]-4-pyrimidinyl]-2-pyridinecarbonitrile |
---|---|
CAS | 675103-36-3 |
Assay Percent Range | >98% |
Molecular Formula | C20H15F3N4O2 |
Formula Weight | Observed MW: 400.35 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18741429
|
Tocris Bioscience
5706/25 |
25 mg |
455.00€
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18739278
|
Tocris Bioscience
5706/5 |
5 mg |
118.00€
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent and selective cell-permeable human neutrophil elastase (HNE) inhibitor (IC50 = 20 nM). Exhibits around 2000-fold selectivity for NHE against a panel of 21 other serine proteases. Effective in an acute lung injury model in vivo. Orally bioavailable. Inactive S-enantiomer (Cat.No.6389) also available.Specifications
5-[(4R)-5-Acetyl-1,2,3,4-tetrahydro-6-methyl-2-oxo-1-[3-(trifluoromethyl)phenyl]-4-pyrimidinyl]-2-pyridinecarbonitrile | |
>98% | |
Observed MW: 400.35 |
675103-36-3 | |
C20H15F3N4O2 |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BAY 678