Learn More
Proteintech ChromoTek Halo-Trap Agarose Beads
Halo-Trap Agarose are affinity beads for IP of Halo-tag fusion proteins
Brand:  Proteintech ota-100
 View more versions of this product
Product Code. 17353403
  • 1798.00€ / 100 reactions
In Stock
Description
  • Comprises a Halo-tag Nanobody /VHH conjugated to agarose beads
Specifications
Specifications
Upon receipt store at 4°C. Do not freeze.
Halo-tag
Alpaca
IP, CoIP, ChIP, RIP
Agarose
100 Reactions