missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek Halo-Trap Agarose Beads
Halo-Trap Agarose are affinity beads for IP of Halo-tag fusion proteins
238.00€ - 5899.00€
Specifications
Content And Storage | Upon receipt store at 4°C. Do not freeze. |
---|---|
For Use With (Application) | IP, CoIP, ChIP, RIP |
Target | Halo-tag |
Label or Dye | Agarose |
Species | Alpaca |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
17343403
|
Proteintech
ota-10 |
10 Reactions |
238.00€
10 reactions |
In Stock
Log in to see stock. |
|||||
17353403
|
Proteintech
ota-100 |
100 Reactions |
1798.00€
100 reactions |
In Stock
Log in to see stock. |
|||||
17363403
|
Proteintech
ota-20 |
20 Reactions |
397.00€
20 reactions |
In Stock
Log in to see stock. |
|||||
17373403
|
Proteintech
ota-200 |
200 Reactions |
3237.00€
200 reactions |
In Stock
Log in to see stock. |
|||||
17383403
|
Proteintech
ota-400 |
400 Reactions |
5899.00€
400 reactions |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
Description
- Comprises a Halo-tag Nanobody /VHH conjugated to agarose beads
Specifications
Upon receipt store at 4°C. Do not freeze. | |
Halo-tag | |
Alpaca |
IP, CoIP, ChIP, RIP | |
Agarose |