Learn More
Proteintech ChromoTek Halo-Trap Agarose Beads
Halo-Trap Agarose are affinity beads for IP of Halo-tag fusion proteins
Brand: Proteintech ota-200
This item is not returnable.
View return policy
Description
- Comprises a Halo-tag Nanobody /VHH conjugated to agarose beads
Specifications
Upon receipt store at 4°C. Do not freeze. | |
Halo-tag | |
Alpaca |
IP, CoIP, ChIP, RIP | |
Agarose | |
200 Reactions |