Learn More
Nabertherm™ Compact Tube Furnace
These compact tube furnaces with integrated control systems can be used universally for many processes. Equipped with a standard working tube of C 530 ceramic and two fiber plugs, these tube furnaces have an unbeatable price/performance ratio.
Brand:  Nabertherm™ R-571C2AN
 View more versions of this product
Product Code. 15426592
Description
Description
  • Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
  • Solid state relays provide for low-noise operation
Specifications
Specifications
C450
1200°C
1400 mm
1000 mm
1170 x 460 x 628 mm
170 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Nabertherm™ Compact Tube Furnace > R 170/1000/12/C450