missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ EIF4EBP1 (Human) Recombinant Protein
Human EIF4EBP1 full-length ORF ( AAH04459, 1 a.a. - 118 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00001978-P01.25ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Sequence: MSGGSSCSQTPSRAIPATRRVVLGDGVQLPPGDYSTTPGGTLFSTTPGGTRIIYDRKFLMECRNSPVTKTPPRDLPTIPGVTSPSSDEPPMEASQSHLRNSPEDKRAGGEESQFEMDI
Especificaciones
AAH04459 | |
eukaryotic translation initiation factor 4E binding protein 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EIF4EBP1 | |
GST |
1978 | |
Wheat germ expression system | |
25 μg | |
4E-BP1, 4EBP1, BP-1, MGC4316, PHAS-I | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |