missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ EIF4EBP1 (Human) Recombinant Protein
Human EIF4EBP1 full-length ORF ( AAH04459, 1 a.a. - 118 a.a.) recombinant protein with GST-tag at N-terminal.
335.00€ - 508.00€
Specifications
Accession Number | AAH04459 |
---|---|
Gene ID (Entrez) | 1978 |
Name | eukaryotic translation initiation factor 4E binding protein 1 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16104441
|
Abnova™
H00001978-P01.10ug |
10 μg |
335.00€
10µg |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||||
16114441
|
Abnova™
H00001978-P01.25ug |
25 μg |
508.00€
25µg |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||||
Description
- Sequence: MSGGSSCSQTPSRAIPATRRVVLGDGVQLPPGDYSTTPGGTLFSTTPGGTRIIYDRKFLMECRNSPVTKTPPRDLPTIPGVTSPSSDEPPMEASQSHLRNSPEDKRAGGEESQFEMDI
Specifications
AAH04459 | |
eukaryotic translation initiation factor 4E binding protein 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
4E-BP1, 4EBP1, BP-1, MGC4316, PHAS-I | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
1978 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EIF4EBP1 | |
GST |
Safety and Handling
missing translation for 'shelfLife' : Best use within three months from the date of receipt of this protein