missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human OSBPL8 Partial ORF (NP_065892.1, 244 a.a. - 346 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00114882-Q01.25ug
This item is not returnable.
View return policy
Description
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: IIRATSESDGRCWMDALELALKCSSLLKRTMIREGKEHDLSVSSDSTHVTFYGLLRANNLHSGDNFQLNDSEIERQHFKDQDMYSDKSDKENDQEHDESDNEVSpecifications
NP_065892.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.07kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
IIRATSESDGRCWMDALELALKCSSLLKRTMIREGKEHDLSVSSDSTHVTFYGLLRANNLHSGDNFQLNDSEIERQHFKDQDMYSDKSDKENDQEHDESDNEV | |
RUO | |
OSBPL8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
114882 | |
OSBPL8 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686A11164/MGC126578/MGC133203/MST120/MSTP120/ORP8/OSBP10 | |
OSBPL8 | |
Recombinant | |
wheat germ expression system |