missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human OSBPL8 Partial ORF (NP_065892.1, 244 a.a. - 346 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_065892.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 114882 |
Molecular Weight (g/mol) | 37.07kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16101447
|
Abnova™
H00114882-Q01.25ug |
25 ug |
508.00€
25µg |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
16191437
|
Abnova™
H00114882-Q01.10ug |
10 ug |
335.00€
10µg |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: IIRATSESDGRCWMDALELALKCSSLLKRTMIREGKEHDLSVSSDSTHVTFYGLLRANNLHSGDNFQLNDSEIERQHFKDQDMYSDKSDKENDQEHDESDNEVSpecifications
NP_065892.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.07kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686A11164/MGC126578/MGC133203/MST120/MSTP120/ORP8/OSBP10 | |
OSBPL8 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
114882 | |
OSBPL8 (Human) Recombinant Protein (Q01) | |
IIRATSESDGRCWMDALELALKCSSLLKRTMIREGKEHDLSVSSDSTHVTFYGLLRANNLHSGDNFQLNDSEIERQHFKDQDMYSDKSDKENDQEHDESDNEV | |
RUO | |
OSBPL8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |