missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CellMaxx™ Bovine Albumin, Low Endotoxin, Stem Cell Culture
Lipid-rich bovine serum albumin, suitable for stem cell culture and media formulation
Brand: MP Biomedicals™ 0219989610
This item is not returnable.
View return policy
Description
Chromatographically purified for culturing embryonic, mesenchymal and hematopoietic cells. Lipid-rich formulation can be substituted for Albumax™ II.
Physical characteristics:
- Form: Off white to brown powder
- Purity:≥98% (w/w)
- Total Protein: ≥96%
- pH (4%): 6.5 to 7.5
- Solubility: Dissolves in 20 minutes at 20° to 25°C
- Moisture: ≤5% w/w
- Endotoxin: ≤1EU/mg
- Mycoplasma: Negative
- Bioburden: ≤100cfu/g
- Virus (9CFR): Negative
- Reported: Lipid content
Certifications: ISO9001, ISO13485, cGMP, EDQM
Specifications
Mammalian Cell Culture | |
66 | |
6.5 to 7.5 | |
10 g | |
4°C |
Lipid-rich | |
CellMaxx™ Bovine Albumin, Low Endotoxin | |
Chromatography | |
Bovine | |
Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction